missing translation for 'onlineSavingsMsg'
Learn More

Resatorvid, MedChemExpress

Product Code. 16465296
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465296 100 mg 100mg
16417127 5 mg 5mg
16404408 10 mg 10mg
16400727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465296 Supplier Medchem Express Supplier No. HY11109

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Resatorvid (TAK-242) is a selective Toll-like receptor 4 (TLR4) inhibitor. Resatorvid inhibits NO, TNF-α and IL-6 production with IC50s of 1.8 nM, 1.9 nM and 1.3 nM, respectively. Resatorvid downregulates expression of TLR4 downstream signaling molecules MyD88 and TRIF. Resatorvid inhibits autophagy and plays pivotal role in various inflammatory diseases.

Resatorvid (TAK-242) is a selective Toll-like receptor 4 (TLR4) inhibitor. Resatorvid inhibits NO, TNF-α and IL-6 production with IC50s of 1.8 nM, 1.9 nM and 1.3 nM, respectively. Resatorvid downregulates expression of TLR4 downstream signaling molecules MyD88 and TRIF. Resatorvid inhibits autophagy and plays pivotal role in various inflammatory diseases.

Specifications

Chemical Name or Material Resatorvid
CAS 243984-11-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H17ClFNO4S
Quantity 100 mg
Synonym TAK-242 CLI-095
Solubility Information DMSO : 100 mg/mL (276.38 mM; Need ultrasonic)
SMILES O=S([C@H]1C(C(OCC)=O)=CCCC1)(NC2=CC=C(C=C2Cl)F)=O
Molecular Weight (g/mol) 361.82
Formula Weight 361.82
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.