missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RERGL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559704
This item is not returnable.
View return policy
Description
Immunogen sequence: CKRAVESAVF LVGNKRDLCH VREVGWEEGQ KLALENRCQF CELSAAEQSL EVEMMFIRII KDILINFKLK EKRRPSGSKS MA Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 79%.
Specifications
| RERGL | |
| Polyclonal | |
| Unconjugated | |
| RERGL | |
| EG632971; ras-related and estrogen-regulated growth inhibitor-like protein; RERG like; RERG/RAS-like; RERG/Ras-like protein; Rergl; RGD1560090 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 500356, 632971, 79785 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H628 | |
| RERGL | |
| Recombinant protein corresponding to Human RERGL. Recombinant protein control fragment (Product #RP-97702). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction