missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RERG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104063
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RERG.
RERG, a member of the RAS superfamily of GTPases, inhibits cell proliferation and tumor formation.
Specifications
| RERG | |
| Polyclonal | |
| Unconjugated | |
| RERG | |
| MGC15754; RAS like estrogen regulated growth inhibitor; RAS-like; RAS-like, estrogen-regulated, growth inhibitor; RAS-like, estrogen-regulated, growth-inhibitor; ras-related and estrogen-regulated growth inhibitor; ras-related and estrogen-regulated growth protein; RERG; RGD1562829; si:cabz01085950.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 232441, 85004 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R367, Q96A58 | |
| RERG | |
| A synthesized peptide derived from human RERG(Accession Q96A58), corresponding to amino acid residues S129-R179. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction