missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RERG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559545
This item is not returnable.
View return policy
Description
Immunogen sequence: TAGQEDTIQR EGHMRWGEGF VLVYDITDRG SFEEVLPLKN ILDEIKKPKN VTLILVGNK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
RERG, a member of the RAS superfamily of GTPases, inhibits cell proliferation and tumor formation.
Specifications
| RERG | |
| Polyclonal | |
| Unconjugated | |
| RERG | |
| MGC15754; RAS like estrogen regulated growth inhibitor; RAS-like; RAS-like, estrogen-regulated, growth inhibitor; RAS-like, estrogen-regulated, growth-inhibitor; ras-related and estrogen-regulated growth inhibitor; ras-related and estrogen-regulated growth protein; RERG; RGD1562829; si:cabz01085950.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 85004 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96A58 | |
| RERG | |
| Recombinant protein corresponding to Human RERG. Recombinant protein control fragment (Product #RP-98443). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction