missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RERE Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94613-0.02ml
This item is not returnable.
View return policy
Description
RERE Polyclonal antibody specifically detects RERE in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RERE | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| ARGARPKIAA0458arginine-glutamic acid dipeptide repeats protein, arginine-glutamic acid dipeptide (RE) repeats, ATN1L, atrophin 2, atrophin-1 like protein, atrophin-1 related protein, Atrophin-1-like protein, Atrophin-1-related protein, DNB1, FLJ38775 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human RERE (NP_036234.3). MTADKDKDKDKEKDRDRDRDREREKRDKARESENSRPRRSCTLEGGAKNYAESDHSEDEDNDNNSATAEESTKKNKKKPPKKKSRYERTDTGEITSYITEDDVVYRPGDCVYIESRRPNTPYFICSIQDFKLVHNSQACCRSPTPALCDPPACSLPVASQPPQHLSEAGRGPVGSKRDHLLMNVKWYYRQSEVPDSVYQHLVQDRHNENDSGRELVITDP | |
| 0.02 mL | |
| Apoptosis, Cell Biology | |
| 473 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction