Learn More
Unity Lab Services™ Centrifuge Standard Requalification Service with Cycle Testing
Product Code. 30178674
Change view
Click to view available options
For Use With (Equipment):
Centrifuge
Unit Size:
1 unit
Product Code. For Use With (Equipment) unitSize
30178674 Centrifuge 1 unit
1 options
Product Code. 30178674 Supplier Unity Lab Services™ Supplier No. RQCTCENSTD
Requalification Services including Cycle testing for Centrifuge Standard (documentation included)
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
  • Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
  • Key tests Min/Max speed, Brief /Extended time, temperature for refrigerated models.
  • Cycle testing (CT): Point-in-time test that monitors a unit and a specified rotor at predefined speed, time, and temperature (if applicable) set points.
  • Testing demonstrates that a unit and specified rotor can meet customer specifications.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Centrifuge
Product Type Requalification Service