missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RepSox (Hydrochloride)

Product Code. 17139251
Encompass_Preferred
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 17139251

Brand: STEMCELL Technologies 72392

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Activin/BMP/TGF-β pathway inhibitor; Inhibits ALK5

  • RepSox is a cell permeable, selective inhibitor of the TGF-β type 1 receptor (TGFβRI) ALK5 (IC50 = 4, 18, and 23 nM for ALK5 autophosphorylation, TGF-β cellular assay, and ALK5 binding in HepG2 cells, respectively; Gellibert et al.) This inhibitor demonstrated less potent activity (IC50 > 16 μM) against 9 related kinases, including p38 MAPK and GSK3 (Gellibert et al.) This product is supplied as the hydrochloride salt of the molecule
  • Enhances reprogramming of mouse embryonic fibroblasts (MEFs) that have been transduced with OCT4, KLF4, and c-MYC (Ichida et al.; Subramanyam et al.)
  • Direct lineage reprogramming of fibroblasts to mature neurons, in combination with CHIR99021, VaLproic Acid, Forskolin, SP600125, Gö6983 and Y-27632 (Hu et al.)
  • Alone or in combination with forskolin, dexamethasone, and nicotinamide, induces differentiation of human pancreatic progenitor cells into insulin-producing cells (Kunisada et al.; Rezania et al.)

Specifications

Synonym Alk 5 Inhibitor II; E 616452; SJN 2511
Quantity 5 mg
Molecular Formula C17H13N5 · HCl
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.