missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REPS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80892-25ul
This item is not returnable.
View return policy
Description
REPS2 Polyclonal specifically detects REPS2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| REPS2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| partner of Ral-binding protein 1, POB1Partner of RalBP1, RALBP1 associated Eps domain containing 2, ralBP1-associated Eps domain-containing protein 2, RalBP1-interacting protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9185 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| REPS2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KCELPLPRFMMSKNDGEIRFGNPAELHGTKVQIPYLTTEKNSFKRMDDEDKQQETQSPTMSPLASPPSSPPHYQRVPLSHGYSKLRSSAEQMHPAPYEARQPLVQPEGSSSGGPGTKPLRHQASLIRSFSVERELQDNSSYPDEPWR | |
| 25ul | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction