Learn More
Sartorius™ ReproEasy Performance Test Pads
GREENER_CHOICE
Product Code. 15753742 Shop All Sartorius Products
Change view
Click to view available options
Type:
Moisture Analyzer Test Pads, 10/Pk.
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
15753742 Moisture Analyzer Test Pads, 10/Pk. Each
15689963 Moisture Analyzer Test Pads, 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15753742 Supplier Sartorius™ Supplier No. YHP02MA
Test pads for the periodic review of the performance of MA37/160 moisture analyzer.
  • Allows you to quickly confirm whether a device is working properly and delivers correct results.
  • Use together with the performance test function in moisture analyzer.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MA37/160 Moisture Analyzer
Quantity 20/Pk.
Type Moisture Analyzer Test Pads, 10/Pk.