missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Repositionable Vinyl Printer Labels

Product Code. 11725683
Change view
Click to view available options
Color:
White
Yellow
Length (Metric):
12.70
12.70 mm
15.24 m
25.40 mm
25.4mm
83.8 m
9.1 m
9.10 m
Width (Metric):
6.1 mm
8.08 mm
9.53 mm
12.70 mm
25.4 mm
38.10 mm
48.26 mm
50.8 mm
Unit Size:
1 roll
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Length (Metric) Width (Metric) unitSize
11725683 White 12.70 38.10 mm Each
12850099 White 12.70 mm 25.4 mm Each
11509463 White 83.8 m 9.53 mm Each
11573984 Yellow 15.24 m 50.8 mm Each
11504064 Yellow 15.24 m 12.70 mm Each
11569463 White 9.1 m 12.70 mm Each
11579463 White 9.1 m 25.4 mm Each
11785663 White 25.40 mm 38.10 mm Each
11563984 White 15.24 m 50.8 mm 1 roll
11594054 White 15.24 m 12.70 mm Each
11765673 White 25.4mm 9.53 mm Each
11578604 White 9.10 m 48.26 mm Each
11549463 White 83.8 m 8.08 mm Each
11508614 White 15.24 m 6.1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 11725683 Supplier Brady™ Supplier No. 114801

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Repositionable vinyl cloth labels for use with Label Printer.

  • Repositionable rubber-based adhesive
  • Semi-gloss finish

Specifications

Description BMP71 Label Printer Labels
Adhesive Type Repositionable
Color White
Finish Semi-gloss
Material Vinyl
Labels per Row 1
Wire Gauge 0
Dimensions (L x W) 12.70 x 38.10 mm
Length (Metric) 12.70
Width (Metric) 38.10 mm
Height (Metric) 12.7 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.