Learn More
MakerBot™ Replicator Z18 Build Plate
Product Code. 16301456
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Replicator Z18 3D Printer
Product Code. For Use With (Equipment)
16301456 Replicator Z18 3D Printer
1 options
Product Code. 16301456 Supplier MakerBot™ Supplier No. MKH000013
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
MakerBot™ Build Plates for the Replicator Z18 3D printer reduce downtime when printing multiple large models.
  • Two parts: Removable plastic top plate and a flat, sturdy aluminum base
  • Makes it easy to neatly remove 3D prints
  • Allows you to move onto your next print while setting aside the most recent print
Specifications
For Use With (Equipment) Replicator Z18 3D Printer
Product Type Build Plate