Learn More
MakerBot™ Replicator Z18 3D Printer
Product Code. 16371446
Encompass_Preferred
Change view
Click to view available options
For Use With: Build plates, build plate tapes, and filaments
Product Code.
16371446 For Use With: Build plates, build plate tapes, and filaments
1 options
Product Code. 16371446 Supplier MakerBot™ Supplier No. MKH000004
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
MakerBot™ Replicator Z18 3D Printer seamlessley integrates with the MakerBot™ 3D EcoSystem of hardware software materials and accessories.
  • Make X-large, ultra tall, professional quality, high resolution prototypes and complex models
  • Print multiple things at once so you can get to work on your next design sooner
  • Enclosed and heated build chamber designed for printing large models with minimal curling
  • Fifth generation technology defines the new standard for ease-of-use, quality, and reliability
  • Powered by the new, user-friendly MakerBot™ Replicator 3D Printing Platform
  • App and cloud enabled
  • Wi-Fi, Ethernet and USB connectivity ensures a seamless production workflow
  • 100-micron layer resolution
  • Choose settings that range from fast draft to finer resolution
Specifications
For Use With (Equipment) Various Build Plates and Filaments
Product Type 3D Printer