Learn More
MakerBot™ Replicator Mini+ Grip Build Surface
Product Code. 16371466
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Replicator Mini+ 3D Printer
Product Code. For Use With (Equipment)
16371466 Replicator Mini+ 3D Printer
1 options
Product Code. 16371466 Supplier MakerBot™ Supplier No. MKH000045
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
MakerBot™ Replicator+ Grip Build Surface offers better adhesion when printing and reduces warping and curling.
  • Provides adhesion when printing
  • Reduced warping and curling
Specifications
For Use With (Equipment) Replicator Mini+ 3D Printer
Product Type Build Plate