Learn More
MakerBot™ Replicator+ Bulid Plate
Product Code. 16341466
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Replicator+ 3D Printer
Product Code. For Use With (Equipment)
16341466 Replicator+ 3D Printer
1 options
Product Code. 16341466 Supplier MakerBot™ Supplier No. MKH000042
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
MakerBot™ Build Plate for the MakerBot™ Replicator+ 3D printer reduces downtime when printing multiple models.
  • Compatible with the MakerBot™ Replicator+
  • Bendable, flex build plate
  • Allows for easy removal of prints
  • Attached on top of it is the grip surface for better adhesion when printing
  • Reduces warping and curling
Specifications
For Use With (Equipment) Replicator+ 3D Printer
Product Type Build Plate