Learn More
MakerBot™ Replicator+ 3D Printer
Product Code. 16331466
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Various Build Plates, Build Plate Tapes, and Filaments
Product Code. For Use With (Equipment)
16331466 Various Build Plates, Build Plate Tapes, and Filaments
1 options
Product Code. 16331466 Supplier MakerBot™ Supplier No. MKH000038
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
MakerBot™ Replicator+ 3D Printer combines unparalleled performance with superior results and lasting reliability.
  • Streamlines desktop 3D printing for professionals and educators
  • Allows you to import native CAD files, remotely manage all connected printers over the Cloud, and optimize prints for speed and quality
  • Approximately 30% faster print time than the Replicator Desktop 3D Printer (5th Generation)
  • Print up to 11.6 x 7.6 x 6.5 in.
  • 25% larger prints than the MakerBot™ Replicator
  • Compatible with Tough PLA Filament Bundle
  • Created so engineers and designers can print durable, high-impact strength prototypes and fixtures
  • On-board camera for remote monitoring with 640 x 480 resolution
  • Connect to the MakerBot Replicator+ using Wi-Fi, USB stick, USB cable or Ethernet
Specifications
For Use With (Equipment) Various Build Plates, Build Plate Tapes, and Filaments
Product Type 3D Printer