missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Replacement Teflon O-rings

Product Code. 17747524 Shop All MP Biomedicals Products
Click to view available options
Packaging Quantity:
50 Pk.
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 17747524

Brand: MP Biomedicals™ 116990100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These Replacement Teflon O-rings are used in the cap of a SS Lysing Matrix tube to provide a leak-proof closure.

  • Replacement o-rings made of teflon designed to be inserted in the cap of the metal Lysing Matrix tubes to provide a leak-proof closure.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 2 mL Metal SS Lysing Matrix Tube
Packaging Quantity 50 Pk.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.