Learn More
Trajan Scientific and Medical™ Replacement SilTite™ Mini Union Ferrules
SilTite Mini Union delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand:  Trajan Scientific and Medical™ 073471
Code : 13
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12947111
  • £73.00 / Pack of 10
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
Specifications
Specifications
Mini Union Ferrules
10/Pk.
0.32 mm