missing translation for 'onlineSavingsMsg'
Learn More

Vapourtec Ltd. Replacement SF-10 Red Pump Tube

Product Code. 15848942
Encompass
Click to view available options
For Use With:
SF-10 Reagent Pump
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15848942

Brand: Vapourtec Ltd. 501300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The innovative SF-10 is a highly chemically resistant pump capable of generating a smooth, continuous flow. The flow rate does not fluctuation like a conventional peristaltic pump offering greater precision. The SF-10 is highly versatile, offering five operating modes (constant flow rate, ramping flow rate, metering gases, volume dispensing and precision pressure regulator) in the one compact unit

  • Replacement red tube for the SF-10 reagent pump

Specifications

For Use With SF-10 Reagent Pump
Color Red
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.