missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vapourtec Ltd. Replacement SF-10 Blue Pump Tube
Click to view available options
For Use With:
SF-10 Reagent Pump
Description
- Replacement blue tube for the SF-10 reagent pump
Specifications
Specifications
| For Use With | SF-10 Reagent Pump |
| Color | Blue |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction