missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Replacement Seals, Pump Assemblies and Valve Block Caps for BRAND seripettor and seripettor pro Bottletop Dispensers

Accessories for BRAND seripettor and seripettor pro Bottletop Dispensers

Brand:  BRAND™ 704542

Additional Details : Weight : 0.07000kg

 View more versions of this product

Product Code. 11376390

  • £69.80 / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

  • 6788 - EPDM seals for seripettor dispensers, pack of 5
  • 704536 - autoclavable filling tube for seripettor, 250mm, each
  • 704538 - autoclavable filling tube for seripettor, 500mm, each
  • 704542 - pump assembly for 1-10mL seripettor
  • 704544 - pump assembly for 2.5-25mL seriipettor
  • 704549 - Pump assembly for 2.5-25mL seripettor pro
  • 704552 - seripettor valve block cap for autoclaving. Fits 1-10mL seripettor
  • 704554 seripettor valve block cap for autoclaving. Fits 2.5-10mL seripettor
Specifications

Specifications

Pump Assembly
Clear, Blue, Red
Seripettor™, 10 mL
Non-sterile
Not Autoclavable
Dispensing liquid
Pump assembly only
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers