missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+
Click to view available options
Type:
Comparator 2000+ Replacement Reagent
Iron HR Tablet
Persulfate Reagent Powder
Silica No. 1 Tablet
Unit Size:
0.5kg
30g
Each
Pack of 100
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
Specifications
| Color | White |
| Composition | Boric Acid 45 to 50%, Calcium Thioglycollate Trihydrate 20 to 25%, Nonhazardous components 10 to 20%, Disodium Tetraborate 5 to 10% |
| pH Range | 8.4 |
| Format | Tablet |
| For Use With (Application) | Water analysis |
| For Use With (Equipment) | Colorimeter and spectrophotometer |
| Quantity | 100/Pk. |
| Range | 1 to 10mg/L |
| Type | Iron HR Tablet |
Safety and Handling
Product Identifier
- iron HR tablets
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : ammonium chloride, calcium thioglycolatetrihydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction