Learn More
Grant Instruments™ Replacement Polycarbonate Transparent Lid, Gabled
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
Brand: Grant Instruments™ AQL2
Code : 57
Additional Details : Weight : 0.10000kg
Description
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
- Polycarbonate construction
- Clear, gabled lid allows condensation buildup to drip away from immersed vessels.
- For use with Sub Aqua Pro Series Unstirred Water Baths
Specifications
Gabled | |
2L Tanks | |
Polycarbonate |