Learn More
BTX™ Replacement Pins for Plate Handler
Product Code. 15447320
Click to view available options
Quantity:
100 Pk.
25 Pk.
Unit Size:
Pack of 100
Pack of 25
Product Code. 15447320
Brand: BTX™ 450468
Replacement Pins designed to work with BTX™ Plate Handlers.
  • Replacement Pins for BTX™ Plate Handlers
Specifications
For Use With (Application) High Throughput transfection
For Use With (Equipment) HT 100, HT 200
Material Gold-plated
Product Line BTX Electroporation
Product Type Electroporation Spare Pin
Description Replacement Pins for Plate Handler, Pkg. of 25
Electrode Shape Pin
Includes 25 x Replacement Pins
Quantity 25 Pk.
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Exponential Decay Wave, Square Wave
High-throughput Compatibility Yes