missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Replacement Parts for C20 Karl Fischer™ Autotitrator

Product Code. 13172152 Shop All Mettler Toledo Products
Change view
Click to view available options
Type:
Coulometric Cell with Diaphragm
Coulometric Cell without Diaphragm
Drying Tube
Septa
Solvent Drain/Delivery Tube
Unit Size:
Each
Pack of 12
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
13172152 Septa Pack of 12
11390532 Coulometric Cell without Diaphragm Each
12655815 Coulometric Cell with Diaphragm Each
11371590 Solvent Drain/Delivery Tube -
13152152 Drying Tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13172152 Supplier Mettler Toledo™ Supplier No. 51108740
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Accessories For use with Mettler Toledo C20 Karl Fischer™ Autotitrator

Coulometric Cells

  • Coulometer cells with or without diaphragm
  • Cell without diaphragm is almost maintenance-free:, innovative design lets you use this cell for determination of water in oils.
  • Cell with a diaphragm is recommended for applications such as the determination of water in substances containing ketones, also recommended for best possible accuracy
Coulometric Reagent Exchange Set
Allows emptying and re-filling the coulometric cell, reducing the risk of the user coming into contact with reagents
TRUSTED_SUSTAINABILITY

Specifications

Type Septa
For Use With (Equipment) C20 Coulometric Titrator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.