Learn More
Mettler Toledo™ Replacement Parts for C20 Karl Fischer™ Autotitrator
Shop All Mettler Toledo Products
Click to view available options
Type:
Coulometric Cell with Diaphragm
Coulometric Cell without Diaphragm
Drying Tube
Septa
Solvent Drain/Delivery Tube
Unit Size:
Each
Pack of 12
Description
Coulometric Cells
- Coulometer cells with or without diaphragm
- Cell without diaphragm is almost maintenance-free:, innovative design lets you use this cell for determination of water in oils.
- Cell with a diaphragm is recommended for applications such as the determination of water in substances containing ketones, also recommended for best possible accuracy
Allows emptying and re-filling the coulometric cell, reducing the risk of the user coming into contact with reagents
Specifications
Specifications
| Type | Drying Tube |
| Material | Glass |