Learn More
Fisherbrand™ Replacement Membrane Kit for Ion Selective Electrode
Allows uptake of the specific ion.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.80000kg
Description
- For Iodide and chloride ISE
Specifications
Replacement Membrane Kit | |
Measuring Iodide ions | |
0 to 14 |