Learn More
WTW™ Replacement Flow Through Vessel
GREENER_CHOICE
Product Code. 10558021
Change view
Click to view available options
For Use With (Equipment):
LR 325/001
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10558021 LR 325/001 Each
1 options
Product Code. 10558021 Supplier WTW™ Supplier No. 302760
Only null left
Add to basket
Add to basket
Perfectly designed to perform measurements without the interference of carbon dioxide through the air or to examine to conductivity of solutions in closed circuits.
  • Overpressure must not exceed 0.5 bar
  • For conductivity measuring cells for applications to prevent air from entering
  • Manufactured from stainless steel, for LR 325/001
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Flow Through Vessel
For Use With (Application) Preventing air entry, closed circuit measurements
Material Stainless Steel
For Use With (Equipment) LR 325/001