missing translation for 'onlineSavingsMsg'
Learn More

Replacement Filter Adapter for RF3000™ Pipette Controller

Product Code. 15746098 Shop All Thermo Scientific CO₂ Incubators Products
Change view
Click to view available options
:
Filter Adapter: For use with: RF3000 Pipette Controller
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15746098 Filter Adapter: For use with: RF3000 Pipette Controller
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15746098 Supplier Heathrow Scientific Supplier No. HS5000RFRFA
Only null left
Add to basket
Edge
Add to basket

A replacement filter adapter for use with the RF3000™ Pipette Controller (12904417) so that users are not tied to a pipette-specific brand, thereby reducing the total cost of ownership.

RF3000™ Pipette Controller

Packed with features for comfortable and accurate pipetting the powerful pump offers rapid, yet sensitive aspiration and dispensing, has adjustable valves and is microprocessor controlled for constant reliability and accuracy.

  • Operates with 100-240 volt electrical supplies worldwide
  • Long-life li-ion battery, with quick charge up to 20 hours continuous use
  • UV resistant housing
  • Universal grip and soft coated triggers with direction indicators, for right or left handed users
  • Ergonomically balanced in the hand with a loaded pipette
  • Bright LEDs indicate three operation modes: high, low, and gravity
  • Toggle operation by pressing the large mode button
  • Five different colored nose cones included to help differentiate department, task or user, as well as a labeling area on the side of the unit
  • Filters are readily available, so you are not tied into a pipette specific brand—helps reduce the total life costs of ownership versus other makes of pipette aids
  • An adapter can be fitted to the nose cone to stabilize small volume and thinner pipettes
  • Low battery indicator; can be recharged when in use

Specifications

Color Natural
For Use With RF3000 Pipette Controller
Material Silicone
Type Filter Adapter
For Use With (Application) Filtered Pipetting
For Use With (Equipment) RF3000™ Pipette Controller (12904417)
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.