missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Replacement Digital Control Panel for Vortex-Genie 2

Product Code. 11695391
Click to view available options
Packaging Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11695391

Brand: Scientific Industries ELA236575

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This Replacement Digital Control Panel is for use with the Digital Vortex-Genie 2 and ABI Digital Vortex-Genie 2 Mixers.

  • Replacement Digital Control Panel is compatible with the 100 V, 120 V, 230 V Digital Vortex-Genie 2 and ABI Digital Vortex-Genie 2 Mixers.

Specifications

Product Type Replacement Keypad
Color Blue
Description Digital Keypad
For Use With (Equipment) Digital Vortex-Genie 2, ABI Digital Vortex-Genie 2 (100 V, 120 V, 230 V)
Packaging Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.