Learn More
Fisherbrand™ Replacement Deuterium Lamp For Shimadzu UV and Visible HPLC Detector
Product Code. 13014120
Change view
Click to view available options
For Use With (Equipment):
Shimadzu HPLC Detector
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
13014120 Shimadzu HPLC Detector Each
1 options
Product Code. 13014120 Supplier Fisherbrand™ Supplier No. 1194126
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 31-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fisherbrand™ Replacement Deuterium Lamp For Shimadzu UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Order Info
Cat. No. 14-500-725 is comparable to Shimadzu part 228-34016-02.
Compliance
WEEE and ROHS compliant
Specifications
Light Source Deuterium
For Use With (Equipment) Shimadzu HPLC Detector
Product Type Replacement Detector Lamp