Learn More
Vici Valco Instruments™ Replacement Cutting Wheel
Product Code. 12782680
Change view
Click to view available options
:
Product Type: Replacement Cutting Wheel
Product Code.
12782680 Product Type: Replacement Cutting Wheel
1 options
Product Code. 12782680 Supplier Vici Valco Instruments™ Supplier No. JR793
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type Replacement Cutting Wheel
For Use With (Application) Cutting 1/16 in. and 1/8 in. SS tubing
For Use With (Equipment) Stainless Steel Tubing Cutter