Learn More
Restek™ Replacement Chemical Traps for Agilent™ Gas Chromatographs
Product Code. p-305482
Encompass_Preferred
Click to view available options
Type:
O-rings for Trap Fittings
Replacement Split Vent Line
Replacement Split Vent Trap
Replacement Trap
Split Vent Trap Assembly
Product Code. 13169722
Brand: Restek™ 23031
Easy to install replacement chemical traps attach to the same fittings as original Agilent GC equipment
  • Built-in frits and adsorbents remove both moisture and hydrocarbons
  • Trap can be regenerated to remove contaminants and restore performance
  • Also available is a Replacement 32 in. Split Line with all installation hardware included
Specifications
Type Split Vent Trap Assembly
For Use With (Application) Gas Chromatography
For Use With (Equipment) Agilent 6850/6890 GCs