Learn More
Restek™ Replacement Chemical Traps for Agilent™ Gas Chromatographs
Product Code. 13179722
Encompass_Preferred
Change view
Click to view available options
Type:
O-rings for Trap Fittings
Replacement Split Vent Line
Replacement Split Vent Trap
Replacement Trap
Split Vent Trap Assembly
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type
13179722 Replacement Trap
13169722 Split Vent Trap Assembly
13141452 O-rings for Trap Fittings
10590863 Replacement Trap
10230713 Replacement Split Vent Trap
13137252 Replacement Split Vent Trap
13147242 Replacement Split Vent Line
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 13179722 Supplier Restek™ Supplier No. 23032
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Easy to install replacement chemical traps attach to the same fittings as original Agilent GC equipment
  • Built-in frits and adsorbents remove both moisture and hydrocarbons
  • Trap can be regenerated to remove contaminants and restore performance
  • Also available is a Replacement 32 in. Split Line with all installation hardware included
Specifications
Type Replacement Trap
For Use With (Application) Gas Chromatography
For Use With (Equipment) Agilent GCs