Learn More
Restek™ Replacement Chemical Traps for Agilent™ Gas Chromatographs
Description
- Built-in frits and adsorbents remove both moisture and hydrocarbons
- Trap can be regenerated to remove contaminants and restore performance
- Also available is a Replacement 32 in. Split Line with all installation hardware included
Specifications
Specifications
| Type | Split Vent Trap Assembly |
| For Use With (Application) | Gas Chromatography |
| For Use With (Equipment) | Agilent 6850/6890 GCs |