Learn More
Restek™ Replacement Chemical Traps for Agilent™ Gas Chromatographs
Click to view available options
Type:
O-rings for Trap Fittings
Replacement Split Vent Line
Replacement Split Vent Trap
Replacement Trap
Split Vent Trap Assembly
Description
- Built-in frits and adsorbents remove both moisture and hydrocarbons
- Trap can be regenerated to remove contaminants and restore performance
- Also available is a Replacement 32 in. Split Line with all installation hardware included
Specifications
Specifications
| Type | Replacement Trap |
| For Use With (Application) | Gas Chromatography |
| For Use With (Equipment) | Agilent GCs |