Learn More
Restek™ Replacement Cables for Agilent GCs
Product Code. p-3186904
Encompass_Preferred
Click to view available options
Type:
HP5890GC Remote Start Cable
HP5890GC to Strip Chart/Integrator Cable
Unit Size:
Each
Product Code. 12982579
Brand: Restek™ 20657
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Replacement cables to connect Agilent 5890 GCs.
Part No. 20652
  • To connect an Agilent 5890 GC to a non-Agilent integrator or standard strip chart recorder
  • Length: 6 ft.
Part No. 20657
  • To connect an Agilent 5890 GC to remote-start another piece of equipment or to start the Agilent 5890 GC from that piece of equipment
  • Length: 6 ft.
Specifications
Type HP5890GC Remote Start Cable
Includes Instructions and wiring diagrams
For Use With Agilent GCs