Learn More
Restek™ Replacement Cables for Agilent GCs
Click to view available options
Type:
HP5890GC Remote Start Cable
HP5890GC to Strip Chart/Integrator Cable
Unit Size:
Each
Description
Part No. 20652
- To connect an Agilent 5890 GC to a non-Agilent integrator or standard strip chart recorder
- Length: 6 ft.
Part No. 20657
- To connect an Agilent 5890 GC to remote-start another piece of equipment or to start the Agilent 5890 GC from that piece of equipment
- Length: 6 ft.
Specifications
Specifications
| Type | HP5890GC Remote Start Cable |
| Includes | Instructions and wiring diagrams |
| For Use With | Agilent GCs |