Learn More
Erlab Molecular Filter - 13-1 - AS
Description
- Molecular filter
- Type of filter: Acid vapours
Specifications
Specifications
| For Use With (Equipment) | Fume Hood |
| Product Type | Carbon Filter |