missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REPIN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557498
This item is not returnable.
View return policy
Description
Immunogen sequence: MLERRCRGPL AMGLAQPRLL SGPSQESPQT LGKESRGLRQ QGTSVAQSGA QAPGRAHRCA HCRRHFPGWV ALWLHTRRCQ ARLPLPCPE Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 74%.
Replication initiator 1 (REPIN1) is a gene encoding a zinc-finger protein that plays a critical role in various metabolic processes. Located on chromosome 7 in humans, REPIN1 is prominently expressed in liver and adipose tissues. It is associated with the regulation of body weight and triglyceride levels, as evidenced by its locus within a quantitative trait locus (QTL) related to these metabolic traits. The REPIN1 protein is involved in enhancing DNA bending, which is crucial for replication initiation. Studies have shown that deleting REPIN1 in the liver can lead to improved insulin sensitivity and altered lipid metabolism, with secondary effects on adipose tissue. Additionally, REPIN1 is implicated in iron metabolism and apoptosis regulation, further underscoring its multifaceted role in cellular processes.
Specifications
| REPIN1 | |
| Polyclonal | |
| Unconjugated | |
| REPIN1 | |
| 60 kDa origin-specific DNA-binding protein; 60 kDa replication initiation region protein; AP4; ATT-binding protein; DHFR oribeta-binding protein RIP60; H_DJ0584D14.12; REPIN1; replication initiation region protein (60kD); replication initiator 1; RIP60; Zfp464; Zinc finger protein 464; zinc finger protein 464 (RIP60); zinc finger protein AP4; ZNF464 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29803 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BWE0 | |
| REPIN1 | |
| Recombinant protein corresponding to Human REPIN1. Recombinant protein control fragment (Product #RP-95769). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction