missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REPIN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35845-100ul
This item is not returnable.
View return policy
Description
REPIN1 Polyclonal antibody specifically detects REPIN1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| REPIN1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| 60 kDa origin-specific DNA-binding protein, 60 kDa replication initiation region protein, ATT-binding protein, DHFR oribeta-binding protein RIP60, H_DJ0584D14.12, replication initiation region protein (60kD), replication initiator 1, RIP60AP4, Zfp464, Zinc finger protein 464, zinc finger protein 464 (RIP60), zinc finger protein AP4, zinc finger proten AP4, ZNF464 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 376-567 of human REPIN1 (NP_037532.2).,, Sequence:, SCDDCGRSFRLERFLRAHQRQHTGERPFTCAECGKNFGKKTHLVAHSRVHSGERPFACEECGRRFSQGSHLAAHRRDHAPDRPFVCPDCGKAFRHKPYLAAHRRIHTGEKPYVCPDCGKAFSQKSNLVSHRRIHTGERPYACPDCDRSFSQKSNLITHRKSHIRDGAFCCAICGQTFDDEERLLAHQKKHDV | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 29803 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction