missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals Repetitive Dispenser
Shop All MP Biomedicals Products
Click to view available options
Capacity (Metric):
0.5 to 2.5 μL Dispensing
50 to 250 μL Dispensing
Unit Size:
1 sheet
Description
- Highly precise (< 1% CV precision).
- Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
- Polypropylene construction.
Specifications
Specifications
| Quantity | 1/Pk. |
| Type | Repetitive Dispenser |
| For Use With (Application) | Repeat, precise liquid dispensing |
| Material | Polypropylene |
| Capacity (Metric) | 0.5 to 2.5 μL Dispensing |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction