missing translation for 'onlineSavingsMsg'
Learn More

Reparixin, MedChemExpress

Product Code. 16466398
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16466398 10 mg 10mg
16453466 25 mg 25mg
16452507 50 mg 50mg
16448987 5 mg 5mg
16420236 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16466398 Supplier Medchem Express Supplier No. HY15251
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Reparixin is a non-competitive allosteric inhibitor of the chemokine receptors CXCR1 and CXCR2 activation with IC50s of 1 and 100 nM, respectively.

Reparixin is a non-competitive allosteric inhibitor of the chemokine receptors CXCR1 and CXCR2 activation with IC50s of 1 and 100 nM, respectively.

Specifications

Chemical Name or Material Reparixin
CAS 266359-83-5
Color White
Purity Grade Notes Research
Molecular Formula C14H21NO3S
Quantity 10 mg
Synonym Repertaxin DF 1681Y
Solubility Information DMSO : ≥ 100 mg/mL (352.87 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CS(=O)(NC([C@@H](C1=CC=C(CC(C)C)C=C1)C)=O)=O
Molecular Weight (g/mol) 283.39
Formula Weight 283.39
Percent Purity 95.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.