missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reparixin L-lysine salt, MedChemExpress
Description
Reparixin L-lysine salt is an allosteric inhibitor of chemokine receptor 1/2 (CXCR1/2) activation.
Specifications
Specifications
| Chemical Name or Material | Reparixin L-lysine salt |
| CAS | 266359-93-7 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H35N3O5S |
| Quantity | 10 mg |
| Synonym | Repertaxin L-lysine salt |
| Solubility Information | H2O : 100 mg/mL (232.79 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (232.79 mM; Need ultrasonic) |
| SMILES | N[C@@H](CCCCN)C(O)=O.CS(=O)(NC([C@@H](C1=CC=C(CC(C)C)C=C1)C)=O)=O |
| Molecular Weight (g/mol) | 429.57 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?