Learn More
BRAND™ Repair Set for Transferpettor™
Product Code. 10130571 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
1, 2, 5 μL
10 μL
100 and 200 μL
20 and 25 μL
50 μL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10130571 50 μL Each
10232511 100 and 200 μL Each
10191771 10 μL Each
15633987 50 μL Each
10726901 20 and 25 μL Each
10538781 1, 2, 5 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10130571 Supplier BRAND™ Supplier No. 701967
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes everything necessary to make small repairs to the BRAND™ Transferpettor™
  • Includes 1 allen key, 1 piston rod with fitted PTFE seal, 1 calibrating gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 PTFE seals
  • Mounting block included for capacities of 20μL and up
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DE-M marked
Volume (Metric) 50 μL
For Use With (Equipment) Transferpettor™
Includes Green mounting block
Quantity 1 Each