missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Repaglinide
GREENER_CHOICE

Product Code. 18054402
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18054402 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18054402 Supplier Tocris Bioscience™ Supplier No. 3805/50
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Kir6 (KATP) channel blocker

Kir6 (KATP) channel blocker that binds with high affinity for SUR1 when co-expressed with Kir6.2 (Kd = 0.42 nM). Antidiabetic glucose regulator with hypoglycaemic effect in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 135062-02-1
Quantity 50 mg
Molecular Formula C27H36N2O4
Formula Weight 452.59
Purity >99%
Molecular Weight (g/mol) 452.60
Chemical Name or Material Repaglinide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.