missing translation for 'onlineSavingsMsg'
Learn More

RENT1/UPF1/hUPF1 Antibody [FITC], Novus Biologicals™

Product Code. 30735871 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735871 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735871 Supplier Novus Biologicals Supplier No. AF5879F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

RENT1/UPF1/hUPF1 Polyclonal antibody specifically detects RENT1/UPF1/hUPF1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RENT1/UPF1/hUPF1
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias ATP-dependent helicase RENT1, EC 3.6.1, EC 3.6.4.-, FLJ43809, HUPF1, KIAA0221FLJ46894, Nonsense mRNA reducing factor 1, NORF1delta helicase, pNORF1, regulator of nonsense transcripts 1, RENT1UP Frameshift 1, UPF1 regulator of nonsense transcripts homolog (yeast), up-frameshift mutation 1 homolog, Up-frameshift suppressor 1 homolog, yeast Upf1p homolog
Host Species Goat
Immunogen E. coli-derived recombinant human RENT1/UPF1, Asp351-Arg600, Accession # NP_002902
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5976
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.