missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RENT1/UPF1/hUPF1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-370AF700
This item is not returnable.
View return policy
Description
RENT1/UPF1/hUPF1 Polyclonal antibody specifically detects RENT1/UPF1/hUPF1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| RENT1/UPF1/hUPF1 | |
| Polyclonal | |
| Western Blot | |
| ATP-dependent helicase RENT1, EC 3.6.1, EC 3.6.4.-, FLJ43809, HUPF1, KIAA0221FLJ46894, Nonsense mRNA reducing factor 1, NORF1delta helicase, pNORF1, regulator of nonsense transcripts 1, RENT1UP Frameshift 1, UPF1 regulator of nonsense transcripts homolog (yeast), up-frameshift mutation 1 homolog, Up-frameshift suppressor 1 homolog, yeast Upf1p homolog | |
| The immunogen recognized by this antibody maps to a region between residue 1075 and the C-terminus (residue 1118) of human Regulator of Nonsense Transcripts 1 (up-frameshift mutation 1 homolog) using the numbering given in entry NP_002902.2 (GeneID 5976). | |
| 0.1 mL | |
| DNA Repair, Transcription Factors and Regulators | |
| 5976 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction