missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Renin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102432
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Renin.
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.
Specifications
| Renin | |
| Polyclonal | |
| Unconjugated | |
| Ren | |
| angiotensin-forming enzyme; angiotensinogenase; aspartyl-protease; D19352; EC 3.4.23; EC 3.4.23.15; FLJ10761; HNFJ2; kidney renin; RATRENAA; Ren; Ren1; Ren-1; Ren1c; Ren1d; Ren-A; RENAA; Renin; renin 1 structural; renin b; renin precursor, renal; renin-1; Rn-1; Rnr | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19701, 24715, 5972 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P00797, P06281, P08424 | |
| Ren, Ren1 | |
| A synthesized peptide derived from human REN(Accession P00797), corresponding to amino acid residues V206-F256. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction