missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Renin ELISA Kit

Product Code. 16312014
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16312014 96 Tests Pack of 96
16322014 10 x 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16312014 Supplier Invitrogen™ Supplier No. BMS2271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Renin Human ELISA Kit quantitates Hu Renin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Renin.

Principle of the method

The Human Renin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00797
Assay Range 93.8 to 6000 pg/mL
Assay Sensitivity 32.6 pg/ml
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates, Plasma, Serum
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5972
Gene Symbol REN
Interassay CV 5.1%
Intraassay CV 3.9
Kit Contents Pre-coated 96 well plate, Standard (2 vials), Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, Streptavidin-HRP, Wash Buffer, Substrate Solution, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 50 μL, Plasma 50 μL, Serum 50 μL
Storage Requirements 2°C to 8°C
Target Renin
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.