missing translation for 'onlineSavingsMsg'
Learn More

Renin Antibody (411525) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30732807 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30732807 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732807 Supplier Novus Biologicals Supplier No. FAB4090CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Renin Monoclonal antibody specifically detects Renin in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Renin
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 411525
Conjugate CoraFluor 1
Formulation PBS
Gene Alias angiotensin-forming enzyme, Angiotensinogenase, EC 3.4.23, EC 3.4.23.15, FLJ10761, HNFJ2, Prorenin, renin, renin precursor, renal
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Renin, Leu24-Arg406, Accession # P00797
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5972
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.